Learn More
Cayman Chemical 5-NItroso-8-quInolInol 50mg
SDP
Catalog No. 502954349
Encompass_Preferred
An HDAC inhibitor, antitumor agent that at 10 μM can induce oxidative stress contributing to apoptosis and differentiation in MCF-7 breast cancer cells; inhibits T. gondii tachyzoite propagation in human fibroblasts (EC50 = 80 nM) and inhibits P. falciparum growth in human red blood cells (EC50 = 1.3 μM)
Catalog No. 50-295-4349 Supplier Cayman Chemical Supplier No. 1385650 MG
Add to Cart
Add to Cart
An HDAC inhibitor, antitumor agent that at 10 μM can induce oxidative stress contributing to apoptosis and differentiation in MCF-7 breast cancer cells; inhibits T. gondii tachyzoite propagation in human fibroblasts (EC50 = 80 nM) and inhibits P. falciparum growth in human red blood cells (EC50 = 1.3 μM)