Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitrothiophene-2-carbonitrile, 98%, Thermo Scientific™

Catalog No. AC449370010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449370010 1 g
AC449370050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449370010 Supplier Thermo Scientific Chemicals Supplier No. 449370010
Add to Cart

Chemical Identifiers

CAS 16689-02-4
Molecular Formula C5H2N2O2S
Molecular Weight (g/mol) 154.15
InChI Key FLYONFCGDKAMIH-UHFFFAOYSA-N
Synonym 5-nitro-2-thiophenecarbonitrile, 5-cyano-2-nitrothiophene, 2-cyano-5-nitrothiophene, 5-nitrothiophene-2-carbonitirile, 5-nitro-thiophene-2-carbonitrile, 2-thiophenecarbonitrile, 5-nitro, zlchem 478, maybridge1_005980, 5-nitro-2-cyanothiophene, acmc-209dv8
PubChem CID 519294
IUPAC Name 5-nitrothiophene-2-carbonitrile
SMILES C1=C(SC(=C1)[N+](=O)[O-])C#N

Specifications

Melting Point 42°C to 46°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 154.15
Percent Purity 98%
Chemical Name or Material 5-Nitrothiophene-2-carbonitrile
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.