Learn More
Enzo Life Sciences Amiloride, 5-(N,N-Dimethyl)-,. hydrochloride (5mg). CAS: 1214-79-5 (free base)
SDP
Supplier:  Enzo Life Sciences ALX550261M005
Encompass_Preferred
Na+/H+ antiporter inhibitor (IC50=6.9 µM) with increased selectivity and potency relative to amiloride (ALX-550-212). Alternative name: 5-(N,N-Dimethyl)amiloride . HCl, DMA . HCl. Purity: ≥97% (TLC). Solubility: Soluble in methanol (10mg/ml) or water (6mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-9513