Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Norbornene-2,3-dicarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. N10295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10295G 5 g
N102925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10295G Supplier TCI America Supplier No. N10295G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3813-52-3
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00003735
InChI Key NIDNOXCRFUCAKQ-UHFFFAOYSA-N
Synonym Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Acid
PubChem CID 97965
IUPAC Name bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
SMILES C1C2C=CC1C(C2C(=O)O)C(=O)O

Specifications

Melting Point 178°C
Color White
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Norbornene-2,3-dicarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.