Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Norbornene-2-Endo-Acetic Acid

Catalog No. 772828250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-282-8250MG 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 77-282-8250MG Supplier Sigma Aldrich Supplier No. 772828250MG
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Carboxylic acid functionalized norbornene. Carboxylic acid can be used to modify amine containing compounds such as lysine residues on proteins/peptides via standard peptide coupling methods. Norbornene has been shown to react with 1,2,4,5-tetrazines in an inverse electron demand Diels-Alder cycloaddition reaction, which is a bioorthogonal click chemistry reaction demonstrated to be useful in biological imaging applications.

Specifications

CAS 2169976-34-3
Molecular Formula C28H38ClN2PPd
Linear Formula C28H38ClN2PPd
MDL Number MFCD22417237
Quantity 2 g
Synonym Bicyclo[2.2.1]hept-5-en-2-ylacetic acid; Norbornene acetic acid
Molecular Weight (g/mol) 575.46 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.