Learn More
Enzo Life Sciences 5' Nucleotidase (5000kU)
SDP
Catalog No. 502011114
Encompass_Preferred
Formulation: Liquid. In TRIS-HCl, pH 7.4, 150mM sodium chloride, 1mM magnesium chloride and 10% (v/v) glycerol. Long Term Storage: -80°C.
Catalog No. 50-201-1114 Supplier Enzo Life Sciences Supplier No. BMLKI3075000
Formulation: Liquid. In TRIS-HCl, pH 7.4, 150mM sodium chloride, 1mM magnesium chloride and 10% (v/v) glycerol. Long Term Storage: -80°C.