Learn More
Crystal Chem Inc 5' Nucleotidase Assay Kit, 96-tests
SDP
Catalog No. 501947676
Encompass_Preferred
The 5’-Nucleotidase Assay Kit is used to quantify levels of 5’-nucleotidase in serum using only a 10 µL sample volume. Highlights: Extended linearity from 0-300 U/L. Stable liquid stable format requires no reagent preparation. Excellent precision with low CV's. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7676 Supplier Crystal Chem Inc Supplier No. 80229
The 5’-Nucleotidase Assay Kit is used to quantify levels of 5’-nucleotidase in serum using only a 10 µL sample volume. Highlights: Extended linearity from 0-300 U/L. Stable liquid stable format requires no reagent preparation. Excellent precision with low CV's. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.