Learn More
Medchemexpress LLC 5 -O- 4 4 -Dimethoxy 10mM 1mL
SDP
Catalog No. 5000205421
Encompass_Preferred
5 -O-(4 4 -Dimethoxytrityl)-2 -deoxyuridine (5 -O-DMT-dU) is a competitive inhibitor of E coli dUTP nucleotidohydrolase (dUTPase) with the Ki higher than 1000 M 5 -O-(4 4 -Dimethoxytrityl)-2 -deoxyuridine can be used in machine-assisted DNA synthesis by synthesizing nucleosidic phosphoramidite blocks[1][2]
Catalog No. 50-002-05421 Supplier Medchemexpress LLC Supplier No. HYW09779210MM/1ML
5 -O-(4 4 -Dimethoxytrityl)-2 -deoxyuridine (5 -O-DMT-dU) is a competitive inhibitor of E coli dUTP nucleotidohydrolase (dUTPase) with the Ki higher than 1000 M 5 -O-(4 4 -Dimethoxytrityl)-2 -deoxyuridine can be used in machine-assisted DNA synthesis by synthesizing nucleosidic phosphoramidite blocks[1][2]