Learn More
Medchemexpress LLC 5'-O-(4,4'-dimethoxytrityl)-2'-O-methyluridine | 103285-22-9 | 98.0% | 560.59 g/mol | C31H32N2O8 | 1 G
SDP
Catalog No. 5000299692
Encompass_Preferred
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine is a DMT-protected uridine nucleoside analog with a 2'-O-methyl ribose modification, used as an intermediate in the chemical synthesis of modified oligonucleotides and nucleoside derivatives. Molecular formula C31H32N2O8; molecular weight 560.59 g/mol. Follow supplier storage recommendations for long-term stability.
  • DMT-protected at the 5'-position for controlled deprotection.
  • 2'-O-methyl modification enhances nuclease resistance.
  • Designed for incorporation into synthetic oligonucleotides.
  • High purity (98.0%) suitable for research applications.
  • Stable as a powder under recommended storage conditions.
Catalog No. 50-002-99692 Supplier Medchemexpress LLC Supplier No. HYW0254381G
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine is a DMT-protected uridine nucleoside analog with a 2'-O-methyl ribose modification, used as an intermediate in the chemical synthesis of modified oligonucleotides and nucleoside derivatives. Molecular formula C31H32N2O8; molecular weight 560.59 g/mol. Follow supplier storage recommendations for long-term stability.
  • DMT-protected at the 5'-position for controlled deprotection.
  • 2'-O-methyl modification enhances nuclease resistance.
  • Designed for incorporation into synthetic oligonucleotides.
  • High purity (98.0%) suitable for research applications.
  • Stable as a powder under recommended storage conditions.