Learn More
Medchemexpress LLC 5'-O-(4,4'-dimethoxytrityl)-2'-O-methyluridine | 103285-22-9 | MFCD00274123 | 98.0% | 560.59 | C31H32N2O8 | 5 G
SDP
Catalog No. 5000301248
Encompass_Preferred
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine is a DMT-protected uridine analog used as a nucleoside building block and intermediate for the synthesis of modified oligonucleotides. It is supplied as a light-yellow solid with defined purity and storage specifications suited for research and preparative oligonucleotide chemistry.
  • DMT protected at the 5' hydroxyl for selective chain assembly.
  • 2'-O-methyl modification supports increased nuclease resistance in oligonucleotides.
  • High purity material suitable for synthesis and analytical workflows.
  • Available in gram-scale pack sizes to accommodate different synthesis scales.
  • Stable under recommended storage conditions to preserve quality over time.
Catalog No. 50-003-01248 Supplier Medchemexpress LLC Supplier No. HYW0254385G
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine is a DMT-protected uridine analog used as a nucleoside building block and intermediate for the synthesis of modified oligonucleotides. It is supplied as a light-yellow solid with defined purity and storage specifications suited for research and preparative oligonucleotide chemistry.
  • DMT protected at the 5' hydroxyl for selective chain assembly.
  • 2'-O-methyl modification supports increased nuclease resistance in oligonucleotides.
  • High purity material suitable for synthesis and analytical workflows.
  • Available in gram-scale pack sizes to accommodate different synthesis scales.
  • Stable under recommended storage conditions to preserve quality over time.