Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5'-O-(4,4'-dimethoxytrityl)-5-methoxyuridine | 2095417-73-3 | 98.2% | 576.59 g·mol⁻¹ | C31H32N2O9 | 10 MG
SDP

Catalog No. 5000285852
Encompass_Preferred

5'-O-(4,4'-Dimethoxytrityl)-5-methoxyuridine is a DMT-protected nucleoside derivative used as a building block in solid-phase oligonucleotide synthesis. The 4,4'-dimethoxytrityl (DMT) group protects the 5'-hydroxyl during stepwise chain assembly, enabling incorporation of 5-methoxyuridine into synthetic oligonucleotides for research applications.

  • DMT-protected 5'-hydroxyl for stepwise synthesis.
  • Designed for incorporation into oligonucleotides.
  • High purity suitable for research use (98.2%).
  • Molecular weight 576.59 g·mol⁻¹.
  • Chemical formula C31H32N2O9.
  • Supplied as a small-scale vial (10 mg).

Catalog No. 50-002-85852 Supplier Medchemexpress LLC Supplier No. HY15416710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5'-O-(4,4'-Dimethoxytrityl)-5-methoxyuridine is a DMT-protected nucleoside derivative used as a building block in solid-phase oligonucleotide synthesis. The 4,4'-dimethoxytrityl (DMT) group protects the 5'-hydroxyl during stepwise chain assembly, enabling incorporation of 5-methoxyuridine into synthetic oligonucleotides for research applications.

  • DMT-protected 5'-hydroxyl for stepwise synthesis.
  • Designed for incorporation into oligonucleotides.
  • High purity suitable for research use (98.2%).
  • Molecular weight 576.59 g·mol⁻¹.
  • Chemical formula C31H32N2O9.
  • Supplied as a small-scale vial (10 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.