Learn More
Medchemexpress LLC 5 -O- Bis 4-methoxyp 1g
SDP
Catalog No. 5000205301
Encompass_Preferred
5 -O-[Bis(4-methoxyphenyl)phenylmethyl]-adenosine is an adenosine analog Adenosine analogs mostly act as smooth muscle vasodilators and have also been shown to inhibit cancer progression Its popular products are adenosine phosphate Acadesine (HY-13417) Clofarabine (HY-A0005) Fludarabine phosphate (HY-B0028) and Vidarabine (HY-B0277)[1]
Catalog No. 50-002-05301 Supplier Medchemexpress LLC Supplier No. HYW0977751G
5 -O-[Bis(4-methoxyphenyl)phenylmethyl]-adenosine is an adenosine analog Adenosine analogs mostly act as smooth muscle vasodilators and have also been shown to inhibit cancer progression Its popular products are adenosine phosphate Acadesine (HY-13417) Clofarabine (HY-A0005) Fludarabine phosphate (HY-B0028) and Vidarabine (HY-B0277)[1]