Learn More
Medchemexpress LLC 5'-O-[bis(4-methoxyphenyl)phenylmethyl]-2'-O-(2-methoxyethyl)-N-(2-methyl-1-oxopropyl)guanosine | 251647-50-4 | MFCD03428520 | 95.0% | 713.78 | C38H43N5O9 | 5 G
SDP
Catalog No. 5000284398
Encompass_Preferred
5'-O-DMT-N2-Ibu-2'-OMe-G is a protected guanosine nucleoside intermediate used in the chemical synthesis of modified oligonucleotides and nucleic acid research. It contains a 5'-O-DMT protecting group, an N2-isobutyryl modification, and a 2'-O-methoxyethyl substitution. Supplied as a purified solid (formula C38H43N5O9; molecular weight 713.78; purity 95.0% by NMR).
  • Protected guanosine intermediate for oligonucleotide synthesis.
  • Contains 5'-O-DMT, N2-isobutyryl, and 2'-O-methoxyethyl groups.
  • High reported purity suitable for research applications.
  • Intended for use as a building block in modified oligonucleotide assembly.
  • Offered in laboratory-friendly premeasured quantities.
Catalog No. 50-002-84398 Supplier Medchemexpress LLC Supplier No. HYW4606665G
5'-O-DMT-N2-Ibu-2'-OMe-G is a protected guanosine nucleoside intermediate used in the chemical synthesis of modified oligonucleotides and nucleic acid research. It contains a 5'-O-DMT protecting group, an N2-isobutyryl modification, and a 2'-O-methoxyethyl substitution. Supplied as a purified solid (formula C38H43N5O9; molecular weight 713.78; purity 95.0% by NMR).
  • Protected guanosine intermediate for oligonucleotide synthesis.
  • Contains 5'-O-DMT, N2-isobutyryl, and 2'-O-methoxyethyl groups.
  • High reported purity suitable for research applications.
  • Intended for use as a building block in modified oligonucleotide assembly.
  • Offered in laboratory-friendly premeasured quantities.