Learn More
Cayman Chemical 5-O-Demethylnobiletin, Purity: ≥98%, CAS: 2174-59-6, 10 mg
SDP
Catalog No. 502913463
Encompass_Preferred
A flavonoid with diverse biological activities; induces neurite outgrowth and the expression of the genes encoding GAP43 and synaptophysin in PC12 cells from 10-20 µM; reduces triglyceride levels in 3T3-L1 preadipocytes from 2.5-20 µM; decreases body weight, intra-abdominal fat, plasma and liver triglyceride levels, and plasma cholesterol levels in a mouse model of high-fat diet-induced obesity at 25 mg/kg; reduces hepatic fibrosis and MDA levels in a mouse model of carbon tetrachloride-induced liver injury; reduces ear edema, inflammatory cell infiltration, and papillar fibrosis in a mouse model of TPA-induced inflammation
Catalog No. 50-291-3463 Supplier Cayman Chemical Supplier No. 3523010 MG
A flavonoid with diverse biological activities; induces neurite outgrowth and the expression of the genes encoding GAP43 and synaptophysin in PC12 cells from 10-20 µM; reduces triglyceride levels in 3T3-L1 preadipocytes from 2.5-20 µM; decreases body weight, intra-abdominal fat, plasma and liver triglyceride levels, and plasma cholesterol levels in a mouse model of high-fat diet-induced obesity at 25 mg/kg; reduces hepatic fibrosis and MDA levels in a mouse model of carbon tetrachloride-induced liver injury; reduces ear edema, inflammatory cell infiltration, and papillar fibrosis in a mouse model of TPA-induced inflammation