Learn More
Medchemexpress LLC 5 -O-DMT-2 -TBDMS-Ur 25g
SDP
Catalog No. 5000206379
Encompass_Preferred
5 -O-DMT-2 -TBDMS-Uridine is a deoxyribonucleoside used for the oligonucleotide synthesis
Catalog No. 50-002-06379 Supplier Medchemexpress LLC Supplier No. 25GHYW102322
5 -O-DMT-2 -TBDMS-Uridine is a deoxyribonucleoside used for the oligonucleotide synthesis