Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5'-O-DMT-3'-O-TBDMS-Ac-rC | 123956-65-0 | 99.2% | 701.88 | 1 ML
SDP

Catalog No. 5000350021
Encompass_Preferred

5'-O-DMT-3'-O-TBDMS-Ac-rC is a modified nucleoside that can be utilized for synthesizing DNA or RNA.

  • Modified nucleoside for DNA or RNA synthesis.
  • Appearance: solid, white to off-white.
  • Storage conditions: 4°C, protected from light; in solvent, -80°C for 6 months; -20°C for 1 month (protect from light).
  • Solubility: 100 mg/mL in DMSO.

Catalog No. 50-003-50021 Supplier Medchemexpress LLC Supplier No. HY13861210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5'-O-DMT-3'-O-TBDMS-Ac-rC is a modified nucleoside that can be utilized for synthesizing DNA or RNA.

  • Modified nucleoside for DNA or RNA synthesis.
  • Appearance: solid, white to off-white.
  • Storage conditions: 4°C, protected from light; in solvent, -80°C for 6 months; -20°C for 1 month (protect from light).
  • Solubility: 100 mg/mL in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.