Learn More
Medchemexpress LLC 5'-O-DMT-5-ethynyl-2'-deoxyuridine 3'-CE phosphoramidite | 188411-06-5 | 97.3% | 917.00 | C50H57N6O9P | 10 MG
SDP
Catalog No. 5000299229
Encompass_Preferred
5'-O-DMT-5-ethynyl-2'-deoxyuridine 3'-CE phosphoramidite is a nucleoside phosphoramidite monomer containing a 5-ethynyl modification on the uridine base and a 3'-cyanoethyl phosphoramidite group. It is formulated for automated solid-phase oligonucleotide synthesis and intended for research use only.
  • Enables incorporation of 5-ethynyl deoxyuridine for post-synthesis click chemistry.
  • Compatible with standard phosphoramidite automated DNA/RNA synthesis workflows.
  • High purity suitable for research applications (approximately 97.3%).
  • White to off-white solid; formula C50H57N6O9P; molecular weight 917.00.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Supplied in milligram quantities for oligonucleotide assembly.
Catalog No. 50-002-99229 Supplier Medchemexpress LLC Supplier No. HY4887310MG
5'-O-DMT-5-ethynyl-2'-deoxyuridine 3'-CE phosphoramidite is a nucleoside phosphoramidite monomer containing a 5-ethynyl modification on the uridine base and a 3'-cyanoethyl phosphoramidite group. It is formulated for automated solid-phase oligonucleotide synthesis and intended for research use only.
  • Enables incorporation of 5-ethynyl deoxyuridine for post-synthesis click chemistry.
  • Compatible with standard phosphoramidite automated DNA/RNA synthesis workflows.
  • High purity suitable for research applications (approximately 97.3%).
  • White to off-white solid; formula C50H57N6O9P; molecular weight 917.00.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Supplied in milligram quantities for oligonucleotide assembly.