Learn More
Medchemexpress LLC 5'-O-DMT-5-Ethynyl-2'-deoxyuridine 3'-CE phosphoramidite | 188411-06-5 | 98.1% | C50H57N6O9P | 100 MG
SDP
Catalog No. 5000358849
Encompass_Preferred
5'-O-DMT-5-Ethynyl-2'-deoxyuridine 3'-CE phosphoramidite is a phosphoramidite monomer. It is used in the synthesis of oligonucleotides and is for research purposes only.
  • Used in the synthesis of oligonucleotides.
  • Functions as a phosphoramidite monomer.
  • Relevant for DNA/RNA synthesis.
  • Related to Cell Cycle/DNA Damage research.
Catalog No. 50-003-58849 Supplier Medchemexpress LLC Supplier No. HY48873100MG
5'-O-DMT-5-Ethynyl-2'-deoxyuridine 3'-CE phosphoramidite is a phosphoramidite monomer. It is used in the synthesis of oligonucleotides and is for research purposes only.
  • Used in the synthesis of oligonucleotides.
  • Functions as a phosphoramidite monomer.
  • Relevant for DNA/RNA synthesis.
  • Related to Cell Cycle/DNA Damage research.