Learn More
Medchemexpress LLC 5'-O-DMT-5-Ethynyl-2'-deoxyuridine 3'-CE phosphoramidite | 188411-06-5 | 98.1% | C50H57N6O9P | 25 MG
SDP
Catalog No. 5000359146
Encompass_Preferred
5'-O-DMT-5-Ethynyl-2'-deoxyuridine 3'-CE phosphoramidite is a phosphoramidite monomer that can be used in the synthesis of oligonucleotides. For research use only.
  • Phosphoramidite monomer
  • Used in oligonucleotide synthesis
  • Suitable for DNA/RNA synthesis research
  • For research use only
Catalog No. 50-003-59146 Supplier Medchemexpress LLC Supplier No. 25MGHY48873
5'-O-DMT-5-Ethynyl-2'-deoxyuridine 3'-CE phosphoramidite is a phosphoramidite monomer that can be used in the synthesis of oligonucleotides. For research use only.
  • Phosphoramidite monomer
  • Used in oligonucleotide synthesis
  • Suitable for DNA/RNA synthesis research
  • For research use only