Learn More
Medchemexpress LLC 5'-O-DMT-5-Ethynyl-2'-deoxyuridine 3'-CE phosphoramidite | 188411-06-5 | 98.1% | C50H57N6O9P | 50 MG
SDP
Supplier:  Medchemexpress LLC HY4887350MG
Encompass_Preferred
5'-O-DMT-5-Ethynyl-2'-deoxyuridine 3'-CE phosphoramidite is a phosphoramidite monomer used in the synthesis of oligonucleotides. This product is a white to off-white solid, suitable for laboratory chemical applications and substance manufacturing.
  • Used in the synthesis of oligonucleotides
  • Functions as a phosphoramidite monomer
  • High purity (98.09%)
  • White to off-white solid appearance
Catalog No. 50-003-58816