Learn More
Medchemexpress LLC 5'-O-DMT-ibu-rG | 81246-83-5 | 95.4% | 655.70 | 1 ML
SDP
Catalog No. 5000404618
Encompass_Preferred
5'-O-DMT-ibu-rG is a useful model for a new class of DNA binding molecules, which can aid in the development of potent and selective anti-cancer agents. It is intended for research use only.
  • Designed for research use only
  • Useful model for DNA binding molecules
  • Aids in the development of anti-cancer agents
  • Appearance: solid, white to off-white
Catalog No. 50-004-04618 Supplier Medchemexpress LLC Supplier No. HY4305810MM/1ML
5'-O-DMT-ibu-rG is a useful model for a new class of DNA binding molecules, which can aid in the development of potent and selective anti-cancer agents. It is intended for research use only.
  • Designed for research use only
  • Useful model for DNA binding molecules
  • Aids in the development of anti-cancer agents
  • Appearance: solid, white to off-white