Learn More
Medchemexpress LLC 5'-O-DMT-N2-Ibu-2'-OMe-G | 251647-50-4 | MFCD03428520 | 98.0% | 713.78 | C38H43N5O9 | 100 MG
SDP
Catalog No. 5000284994
Encompass_Preferred
Protected guanosine nucleoside intermediate used in the synthesis of antisense oligonucleotides. It is used to build sequence-specific oligonucleotides that bind complementary mRNA, blocking translation and modulating gene expression for research into genetic diseases and oncology. Supplied as a powder for research use only.
  • Protected nucleoside intermediate for antisense oligonucleotide synthesis.
  • Blocks mRNA translation to modulate gene expression.
  • High purity suitable for research (98.0%).
  • Powder form; recommended storage at 4 °C under nitrogen.
  • Molecular formula C38H43N5O9 and molecular weight 713.78.
Catalog No. 50-002-84994 Supplier Medchemexpress LLC Supplier No. HYW460666100MG
Protected guanosine nucleoside intermediate used in the synthesis of antisense oligonucleotides. It is used to build sequence-specific oligonucleotides that bind complementary mRNA, blocking translation and modulating gene expression for research into genetic diseases and oncology. Supplied as a powder for research use only.
  • Protected nucleoside intermediate for antisense oligonucleotide synthesis.
  • Blocks mRNA translation to modulate gene expression.
  • High purity suitable for research (98.0%).
  • Powder form; recommended storage at 4 °C under nitrogen.
  • Molecular formula C38H43N5O9 and molecular weight 713.78.