Learn More
Medchemexpress LLC 5'-O-DMT-N2-Ibu-2'-OMe-G | 251647-50-4 | MFCD03428520 | 98.0% | 713.78 | C38H43N5O9 | 250 MG
SDP
Catalog No. 5000284992
Encompass_Preferred
5'-O-DMT-N2-Ibu-2'-OMe-G is a DMT-protected guanosine derivative used as an intermediate in the synthesis of antisense oligonucleotides. It contains a 5′-O-DMT protecting group, an N2-isobutyryl modification, and a 2'-O-methoxyethyl substitution that facilitate incorporation into oligonucleotide sequences for research applications.
  • Used as an intermediate for antisense oligonucleotide synthesis.
  • Contains 5′-O-DMT protection for stepwise oligonucleotide assembly.
  • Features N2-isobutyryl and 2'-O-methoxyethyl modifications to enhance stability and specificity.
  • Reported purity suitable for research use (approximately 98%).
  • Supplied as a research chemical for laboratory applications only.
Catalog No. 50-002-84992 Supplier Medchemexpress LLC Supplier No. HYW460666250MG
5'-O-DMT-N2-Ibu-2'-OMe-G is a DMT-protected guanosine derivative used as an intermediate in the synthesis of antisense oligonucleotides. It contains a 5′-O-DMT protecting group, an N2-isobutyryl modification, and a 2'-O-methoxyethyl substitution that facilitate incorporation into oligonucleotide sequences for research applications.
  • Used as an intermediate for antisense oligonucleotide synthesis.
  • Contains 5′-O-DMT protection for stepwise oligonucleotide assembly.
  • Features N2-isobutyryl and 2'-O-methoxyethyl modifications to enhance stability and specificity.
  • Reported purity suitable for research use (approximately 98%).
  • Supplied as a research chemical for laboratory applications only.