Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5'-O-DMT-N2-Ibu-2'-OMe-G | 251647-50-4 | MFCD03428520 | 98.0% | 713.78 g/mol | C38H43N5O9 | 1 G
SDP

Catalog No. 5000284396
Encompass_Preferred

5'-O-DMT-N2-Ibu-2'-OMe-G is a protected guanosine derivative used as an intermediate in the synthesis of antisense oligonucleotides and related nucleic acid chemistries. It is supplied for research use and characterized for synthetic applications.

  • Protected guanosine derivative for oligonucleotide synthesis.
  • Reported purity 98.0%.
  • Molecular weight 713.78 g/mol.
  • Molecular formula C38H43N5O9.
  • Available in laboratory-scale package sizes, including 1 G.
  • Intended as a drug intermediate and reference compound for synthesis.

Catalog No. 50-002-84396 Supplier Medchemexpress LLC Supplier No. HYW4606661G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5'-O-DMT-N2-Ibu-2'-OMe-G is a protected guanosine derivative used as an intermediate in the synthesis of antisense oligonucleotides and related nucleic acid chemistries. It is supplied for research use and characterized for synthetic applications.

  • Protected guanosine derivative for oligonucleotide synthesis.
  • Reported purity 98.0%.
  • Molecular weight 713.78 g/mol.
  • Molecular formula C38H43N5O9.
  • Available in laboratory-scale package sizes, including 1 G.
  • Intended as a drug intermediate and reference compound for synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.