Learn More
Medchemexpress LLC 5'-O-DMT-N4-Ac-2'-F-dC | 159414-98-9 | 99.1% | 589.61 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13860210MM/1ML
Encompass_Preferred
5'-O-DMT-N4-Ac-2'-F-dC is a modified nucleotide commonly used in the synthesis of modified oligonucleotides. Oligonucleotides are short-chain nucleic acids that can "silence" specific genes by pairing with DNA, mRNA, or pre-mRNA.
  • Modified nucleotide for oligonucleotide synthesis
  • Used in DNA/RNA synthesis research
  • Acts as a nucleoside antimetabolite/analog
  • Potential applications in cancer research (e.g., blood diseases, lung cancer, leukemia)
  • Relevant to cell cycle/DNA damage studies
  • Exhibits inhibitor properties
  • For research use only
Catalog No. 50-003-49844