Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5'-O-DMT-N4-Ac-2'-F-dC | 159414-98-9 | 99.1% | 589.61 | 100 MG
SDP

Catalog No. 5000350849
Encompass_Preferred

This modified nucleotide is commonly used in the synthesis of modified oligonucleotides and is intended for research use only. It is presented as a white to off-white solid with a purity of 99.11% and a molecular weight of 589.61.

  • Used in the synthesis of modified oligonucleotides
  • Solid form for stability
  • Purity of 99.11%
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-003-50849 Supplier Medchemexpress LLC Supplier No. HY138602100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This modified nucleotide is commonly used in the synthesis of modified oligonucleotides and is intended for research use only. It is presented as a white to off-white solid with a purity of 99.11% and a molecular weight of 589.61.

  • Used in the synthesis of modified oligonucleotides
  • Solid form for stability
  • Purity of 99.11%
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.