Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5'-O-DMT-N4-Ac-2'-F-dC | 159414-98-9 | 99.1% | 589.61 | 500 MG
SDP

Catalog No. 5000350852
Encompass_Preferred

5'-O-DMT-N4-Ac-2'-F-dC is a modified nucleotide commonly used in the synthesis of modified oligonucleotides. It is for research use only and not sold to patients.

  • Solid appearance
  • White to off-white color
  • Molecular weight of 589.61
  • Formula: C32H32FN3O7
  • Soluble in DMSO (250 mg/mL)
  • Shipping at room temperature
  • Store solid at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
  • Used in research for gene silencing

Catalog No. 50-003-50852 Supplier Medchemexpress LLC Supplier No. HY138602500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5'-O-DMT-N4-Ac-2'-F-dC is a modified nucleotide commonly used in the synthesis of modified oligonucleotides. It is for research use only and not sold to patients.

  • Solid appearance
  • White to off-white color
  • Molecular weight of 589.61
  • Formula: C32H32FN3O7
  • Soluble in DMSO (250 mg/mL)
  • Shipping at room temperature
  • Store solid at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
  • Used in research for gene silencing

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.