Learn More
Medchemexpress LLC 5'-O-DMT-N4-Bz-2'-F-dC | 146954-77-0 | 99.9% | 651.68 | 1 ML
SDP
Catalog No. 5000348919
Encompass_Preferred
5'-O-DMT-N4-Bz-2'-F-dC is a nucleoside that offers protective and modification effects. This product is a white to off-white solid, intended for research use only.
  • Nucleoside with protective and modification effects
  • White to off-white solid
  • Intended for research use only
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month, protected from light
Catalog No. 50-003-48919 Supplier Medchemexpress LLC Supplier No. HY13860310MM/1ML
5'-O-DMT-N4-Bz-2'-F-dC is a nucleoside that offers protective and modification effects. This product is a white to off-white solid, intended for research use only.
  • Nucleoside with protective and modification effects
  • White to off-white solid
  • Intended for research use only
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month, protected from light