Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5'-O-DMT-N4-Bz-2'-F-dC | 146954-77-0 | 99.9% | 651.68 | 500 MG
SDP

Catalog No. 5000350836
Encompass_Preferred

5'-O-DMT-N4-Bz-2'-F-dC is a nucleoside with protective and modification effects, intended for research use only.

  • Offers protective and modification effects
  • Intended for research use only
  • Purity of 99.85%
  • Molecular weight of 651.68
  • Available as a white to off-white solid
  • Soluble in DMSO

Catalog No. 50-003-50836 Supplier Medchemexpress LLC Supplier No. HY138603500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5'-O-DMT-N4-Bz-2'-F-dC is a nucleoside with protective and modification effects, intended for research use only.

  • Offers protective and modification effects
  • Intended for research use only
  • Purity of 99.85%
  • Molecular weight of 651.68
  • Available as a white to off-white solid
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.