Learn More
Medchemexpress LLC 5'-O-DMT-N6-ibu-dA | 190834-10-7 | 98.0% | 623.70 | 1 ML
SDP
Catalog No. 5000352454
Encompass_Preferred
5'-O-DMT-N6-ibu-dA is a nucleoside antimetabolite/analog used in the synthesis of oligodeoxyribonucleotides. It targets DNA/RNA Synthesis and the Cell Cycle/DNA Damage pathway.
  • Can be used in the synthesis of oligodeoxyribonucleotides
  • Store at 4°C, sealed, away from moisture and light
  • When in solvent, store at -80°C for up to 6 months, or -20°C for up to 1 month, sealed, away from moisture and light
Catalog No. 50-003-52454 Supplier Medchemexpress LLC Supplier No. HY13860010MM/1ML
5'-O-DMT-N6-ibu-dA is a nucleoside antimetabolite/analog used in the synthesis of oligodeoxyribonucleotides. It targets DNA/RNA Synthesis and the Cell Cycle/DNA Damage pathway.
  • Can be used in the synthesis of oligodeoxyribonucleotides
  • Store at 4°C, sealed, away from moisture and light
  • When in solvent, store at -80°C for up to 6 months, or -20°C for up to 1 month, sealed, away from moisture and light