Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-O-(E)-p-Coumaroylquinic acid | 5746-55-4 | 338.31 | 5 MG
SDP

Catalog No. 5000415932
Encompass_Preferred

5-O-(E)-p-Coumaroylquinic acid, a quinic acid derivative, is a potent phytochemical agent against hepatitis B virus. For research use only; not for sale to patients.

  • Reduces HBV DNA level in the release of mature HBV particles from HepG2.2.15 cultivation.
  • Purity: 99.86%.
  • Molecular weight: 338.31.
  • Formula: C16H18O8.
  • CAS No.: 5746-55-4.
  • Appearance: Solid.
  • Color: White to off-white.
  • Initial source: Plants (Leguminosae Spatholobus suberectus Dunn, Compositae).

Catalog No. 50-004-15932 Supplier Medchemexpress LLC Supplier No. HYN105435MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-O-(E)-p-Coumaroylquinic acid, a quinic acid derivative, is a potent phytochemical agent against hepatitis B virus. For research use only; not for sale to patients.

  • Reduces HBV DNA level in the release of mature HBV particles from HepG2.2.15 cultivation.
  • Purity: 99.86%.
  • Molecular weight: 338.31.
  • Formula: C16H18O8.
  • CAS No.: 5746-55-4.
  • Appearance: Solid.
  • Color: White to off-white.
  • Initial source: Plants (Leguminosae Spatholobus suberectus Dunn, Compositae).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.