Learn More
Medchemexpress LLC 5'-O-(L-leucylsulfamoyl)adenosine | 288591-93-5 | MFCD31619266 | 97.0% | 459.5 g/mol | C16H25N7O7S | 5 MG
SDP
Catalog No. 5000264570
Encompass_Preferred
Leu-AMS is a leucine analogue small molecule that potently inhibits leucyl-tRNA synthetase (LRS). It exhibits low-nanomolar inhibitory activity and is used as a research reagent for biochemical, cellular, and antimicrobial studies.
  • Potent leucyl-tRNA synthetase inhibitor (IC50 ≈ 22.34 nM).
  • Leucine analogue useful for mechanistic studies of aminoacyl-tRNA synthetase function.
  • Demonstrated cytotoxicity in cancer and normal cell lines.
  • Reported antibacterial activity in growth inhibition assays.
  • Provided for research applications in biochemical and cellular experiments.
Catalog No. 50-002-64570 Supplier Medchemexpress LLC Supplier No. HY1089005MG
Leu-AMS is a leucine analogue small molecule that potently inhibits leucyl-tRNA synthetase (LRS). It exhibits low-nanomolar inhibitory activity and is used as a research reagent for biochemical, cellular, and antimicrobial studies.
  • Potent leucyl-tRNA synthetase inhibitor (IC50 ≈ 22.34 nM).
  • Leucine analogue useful for mechanistic studies of aminoacyl-tRNA synthetase function.
  • Demonstrated cytotoxicity in cancer and normal cell lines.
  • Reported antibacterial activity in growth inhibition assays.
  • Provided for research applications in biochemical and cellular experiments.