Learn More
Cayman Chemical 5-O-Methylvisammioside, Purity: ≥98%, CAS: 84272-85-5, 50 mg
SDP
Catalog No. 502917206
Encompass_Preferred
A chromone with diverse biological activities; inhibits the protein-protein interaction between histone H3 and 14-3-3ε and phosphorylation of histone H3, as well as induces cell cycle arrest at the G2/M phase, in HT-29 cells at 10 µM; inhibits compound 48/80-induced histamine release in LAD 2 mast cells from 10-100 µM; inhibits perivascular adventitia leukocyte infiltration and improves neurological deficits in a rat model of subarachnoid hemorrhage-induced vasospasm at 400 µg/kg; has been used as a quality control marker of S. diviaricata roots
Catalog No. 50-291-7206 Supplier Cayman Chemical Supplier No. 3925050 MG
A chromone with diverse biological activities; inhibits the protein-protein interaction between histone H3 and 14-3-3ε and phosphorylation of histone H3, as well as induces cell cycle arrest at the G2/M phase, in HT-29 cells at 10 µM; inhibits compound 48/80-induced histamine release in LAD 2 mast cells from 10-100 µM; inhibits perivascular adventitia leukocyte infiltration and improves neurological deficits in a rat model of subarachnoid hemorrhage-induced vasospasm at 400 µg/kg; has been used as a quality control marker of S. diviaricata roots