Learn More
Medchemexpress LLC 5-O-Methylvisammioside | 84272-85-5 | 452.45 | 100 MG
SDP
Catalog No. 5000388908
Encompass_Preferred
5-O-Methylvisammioside is a natural compound isolated from *Saposhnikovia divaricata*.
  • 99.90% purity
  • White to off-white solid appearance
  • Soluble in DMSO and H2O for in vitro use
  • Compatible with various in vivo dissolution protocols
Catalog No. 50-003-88908 Supplier Medchemexpress LLC Supplier No. HYN0442100MG
5-O-Methylvisammioside is a natural compound isolated from *Saposhnikovia divaricata*.
  • 99.90% purity
  • White to off-white solid appearance
  • Soluble in DMSO and H2O for in vitro use
  • Compatible with various in vivo dissolution protocols