Learn More
Medchemexpress LLC 5'-O-TBDMS-Bz-dA | 51549-39-4 | 98.37% | 469.61 | 1 ML
SDP
Catalog No. 5000347871
Encompass_Preferred
5'-O-TBDMS-Bz-dA is a nucleoside that acts as a protective and modification agent. This white to off-white solid is for research purposes only, not for human use.
  • Protective and modification effects
  • Used in DNA/RNA synthesis research
  • Classified as a nucleoside analog and adenosine derivative
  • Related to cancer, blood, and cardio-cerebrovascular disease research
  • Soluble in DMSO: 100 mg/mL
  • Formulatable for in vivo experiments
Catalog No. 50-003-47871 Supplier Medchemexpress LLC Supplier No. HY13859510MM/1ML
5'-O-TBDMS-Bz-dA is a nucleoside that acts as a protective and modification agent. This white to off-white solid is for research purposes only, not for human use.
  • Protective and modification effects
  • Used in DNA/RNA synthesis research
  • Classified as a nucleoside analog and adenosine derivative
  • Related to cancer, blood, and cardio-cerebrovascular disease research
  • Soluble in DMSO: 100 mg/mL
  • Formulatable for in vivo experiments