Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-O-TBDMS-N4-Benzoyl-2-deoxycytidine | 51549-36-1 | 98.0% | 445.58 | 10 MM X 1 ML
SDP

Catalog No. 5000350835
Encompass_Preferred

5-O-TBDMS-N4-Benzoyl-2-deoxycytidine is a modified nucleoside suitable for use in the synthesis of deoxyribonucleic acid or nucleic acid.

  • Modified nucleoside for various research applications
  • Purity of 98.0%
  • Available as a white to off-white solid
  • Stable under recommended storage conditions
  • Soluble in DMSO at 250 mg/mL

Catalog No. 50-003-50835 Supplier Medchemexpress LLC Supplier No. HY13859310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-O-TBDMS-N4-Benzoyl-2-deoxycytidine is a modified nucleoside suitable for use in the synthesis of deoxyribonucleic acid or nucleic acid.

  • Modified nucleoside for various research applications
  • Purity of 98.0%
  • Available as a white to off-white solid
  • Stable under recommended storage conditions
  • Soluble in DMSO at 250 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.