Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-O-tert-Butyldimethylsilyl-2,3-O-isopropylidene-alpha,beta-D-ribofuranose, Thermo Scientific™

Catalog No. AAL15986MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15986MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL15986MD Supplier Thermo Scientific Chemicals Supplier No. L15986MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-O-tert-Butyldimethylsilyl-2,3-O-isopropylidene-α,β-D-ribofuranose is used as a starting material for the preparation of N-benzyl alfa-L-manno aminocyclopentitol , 4-deoxy-N-benzyl-alfa-L-manno aminocyclopentitol and N-benzyl 6,7-di-epi-trehalamine using stereoselective allylation. It is also used in oligosaccharides.

Solubility
Soluble in dichloromethane and ethyl acetate.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68703-51-5
Molecular Formula C14H28O5Si
Molecular Weight (g/mol) 304.458
MDL Number MFCD01075716
InChI Key JJLRGSYXWAKGJM-SGUBAKSOSA-N
Synonym 5-o-tert-butyldimethylsilyl-2,3-o-isopropylidene-d-ribose, 3as,4r,6r-6-tert-butyldimethylsilyl oxy methyl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-ol
PubChem CID 71314364
IUPAC Name (3aS,4R,6R)-6-[[tert-butyl(dimethyl)silyl]oxymethyl]-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
SMILES CC1(OC2C(OC(C2O1)O)CO[Si](C)(C)C(C)(C)C)C

Specifications

Melting Point 50°C to 53°C
Quantity 250 mg
Solubility Information Soluble in dichloromethane and ethyl acetate.
Optical Rotation −13.3° (c=1 in Chloroform)
Formula Weight 304.46
Chemical Name or Material 5-O-tert-Butyldimethylsilyl-2,3-O-isopropylidene-alpha,beta-D-ribofuranose
TSCA No
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.