Learn More
Cayman Chemical 5-OAHSA-d17 100ug
SDP
Catalog No. 502979596
Encompass_Preferred
An internal standard for the quantification of 5-OAHSA by GC- or LC-MS
Catalog No. 50-297-9596 Supplier Cayman Chemical Supplier No. 17198100 UG
An internal standard for the quantification of 5-OAHSA by GC- or LC-MS