Learn More
Cayman Chemical 5-Octyl D-glutamate, Purity: ≥98%, CAS: 149332-65-0, 10 mg
SDP
Catalog No. 502973181
Encompass_Preferred
A cell-permeable form of D-glutamate; generates free D-glutamate upon hydrolysis of the ester bond by cytoplasmic esterases; may be used to increase levels of cytoplasmic D-glutamate
Catalog No. 50-297-3181 Supplier Cayman Chemical Supplier No. 1610610 MG
A cell-permeable form of D-glutamate; generates free D-glutamate upon hydrolysis of the ester bond by cytoplasmic esterases; may be used to increase levels of cytoplasmic D-glutamate