Learn More
Cayman Chemical 5-Octyl D-glutamate, Purity: ≥98%, CAS: 149332-65-0, 5 mg
SDP
Catalog No. 502971170
Encompass_Preferred
A cell-permeable form of D-glutamate; generates free D-glutamate upon hydrolysis of the ester bond by cytoplasmic esterases; may be used to increase levels of cytoplasmic D-glutamate
Catalog No. 50-297-1170 Supplier Cayman Chemical Supplier No. 161065 MG
A cell-permeable form of D-glutamate; generates free D-glutamate upon hydrolysis of the ester bond by cytoplasmic esterases; may be used to increase levels of cytoplasmic D-glutamate