Learn More
Cayman Chemical 5-Octyl L-glutamate, Purity: ≥95%, CAS: 4852-91-9, 5 mg
SDP
Catalog No. 502972113
Encompass_Preferred
A stable, cell-permeable molecule which generates free glutamic acid upon hydrolysis of the ester bond by cytoplasmic esterases; used in experiments to increase levels of cytoplasmic glutamic acid; also used to synthesize 5-octyl ester derivatives, including 5-octyl α-ketoglutarate, for delivery to cells
Catalog No. 50-297-2113 Supplier Cayman Chemical Supplier No. 161075 MG
A stable, cell-permeable molecule which generates free glutamic acid upon hydrolysis of the ester bond by cytoplasmic esterases; used in experiments to increase levels of cytoplasmic glutamic acid; also used to synthesize 5-octyl ester derivatives, including 5-octyl α-ketoglutarate, for delivery to cells