Learn More
Medchemexpress LLC 5'-ODMT cEt G phosphoramidite (amidite) | 945628-66-0 | 99.1% | 881.95 | 100 MG
SDP
Catalog No. 5000361354
Encompass_Preferred
5'-ODMT cEt G Phosphoramidite amidite is a potent nucleic acid analog. It shows excellent safety and antisense activity.
  • Enhances the RNA-binding ability.
  • Affects the construction of antisense oligonucleotides (ASOs).
  • Reduces the toxicity of ASOs through alternation of nucleotides conformation and 5'-alkyl DNA modification.
Catalog No. 50-003-61354 Supplier Medchemexpress LLC Supplier No. HY148504100MG
5'-ODMT cEt G Phosphoramidite amidite is a potent nucleic acid analog. It shows excellent safety and antisense activity.
  • Enhances the RNA-binding ability.
  • Affects the construction of antisense oligonucleotides (ASOs).
  • Reduces the toxicity of ASOs through alternation of nucleotides conformation and 5'-alkyl DNA modification.