Learn More
Cayman Chemical 5-OxoETE methyl ester 100ug
SDP
Catalog No. 502933386
Encompass_Preferred
An esterified form of 5-oxoETE; an agonist of OXER1 (EC50 = 1.54 µM) that has a higher maximal response than 5-oxoETE in a β-arrestin assay
Catalog No. 50-293-3386 Supplier Cayman Chemical Supplier No. 26059100 UG
An esterified form of 5-oxoETE; an agonist of OXER1 (EC50 = 1.54 µM) that has a higher maximal response than 5-oxoETE in a β-arrestin assay