Learn More
Cliawaived Inc IDTC-II 5 Panel Instant Drug Test Cards
SDP
Catalog No. NC1859236
Encompass
(25 Tests/Box)Tests for BZO300 / COC150 / MET500 / OPI (MOR 300) / THC50The newly designed IDTC-II dip card features a hermetic seal to prevent the card from flooding. The device features a flat and separate cap so it can be easily photographed without contamination.
This item is not returnable. View return policy
Catalog No. NC1859236 Supplier Cliawaived Inc Supplier No. CLIAIDTCII053571
(25 Tests/Box)Tests for BZO300 / COC150 / MET500 / OPI (MOR 300) / THC50The newly designed IDTC-II dip card features a hermetic seal to prevent the card from flooding. The device features a flat and separate cap so it can be easily photographed without contamination.