Learn More
Cliawaived Inc CLIAwaived, Inc. 5 Panel Cups - IDTC II
SDP
Catalog No. NC1859231
Encompass_Preferred
(25 Cups)CLIAwaived, Inc. - INSTANT DRUG TEST CUP/CARD II (IDTC II). This 5-panel, no-leak drug test cup tests for COC, MET, THC, OPI (MOR 300) and OXY. Results in 5 minutes. FDA-cleared and CLIA-waived.
This item is not returnable. View return policy
Catalog No. NC1859231 Supplier Cliawaived Inc Supplier No. CLIAIDTC560527A
(25 Cups)CLIAwaived, Inc. - INSTANT DRUG TEST CUP/CARD II (IDTC II). This 5-panel, no-leak drug test cup tests for COC, MET, THC, OPI (MOR 300) and OXY. Results in 5 minutes. FDA-cleared and CLIA-waived.