Learn More
Cayman Chemical 5-pApAsodIum salt 700ug
SDP
Catalog No. 502948406
Encompass_Preferred
A linearized form of cyclic di-AMP; intended for use as a negative control for cyclic di-AMP signaling
Catalog No. 50-294-8406 Supplier Cayman Chemical Supplier No. 33886700 UG
A linearized form of cyclic di-AMP; intended for use as a negative control for cyclic di-AMP signaling