Learn More
BTNX Inc 5 Parameter Urine Drug Screen - 25/kit (5.1) - COC300 AMP1000 MET1000 THC50 OPI2000
Catalog No. NC3361247
Encompass_Preferred
The Rapid Response™ Multi-Drug One Step Cup is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs in urine samples. This format is a great alternative to the industry's complicated testing processes and is available with and without adulterant tests. It has a longer detection window than saliva tests and provides results in 5 minutes. This product is also available with an option of built-in Specimen Validity/Adulteration Test Strips.   (5.1) - COC300 AMP1000 MET1000 THC50 OPI2000
This item is not returnable. View return policy
Catalog No. NC3361247 Supplier BTNX Inc Supplier No. D5.11V
The Rapid Response™ Multi-Drug One Step Cup is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs in urine samples. This format is a great alternative to the industry's complicated testing processes and is available with and without adulterant tests. It has a longer detection window than saliva tests and provides results in 5 minutes. This product is also available with an option of built-in Specimen Validity/Adulteration Test Strips.   (5.1) - COC300 AMP1000 MET1000 THC50 OPI2000