Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5 Percent Ruthenium on Activated Carbon Powder, Reduced, Water Wet, 27 microns, BASF Catalyst, Spectrum™ Chemical
SDP

Catalog No. 18607973 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-607-973 5 g
18-607-974 25 g
18-607-975 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-973 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SP4005G
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
  • Free flowing powder with particle size of 27 microns

Chemical Identifiers

CAS 7440-18-8 , 7732-18-5
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
IUPAC Name ruthenium
SMILES [Ru]

Specifications

Physical Form Solid
Quantity 5 g
Solubility Information Insoluble in Water
Concentration or Composition (by Analyte or Components) 5% Ruthenium, 45% Charcoal activated, 50% Water
Odor Odorless
Density 1.8 to 2.1 g/cm³

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.