Learn More
Cayman Chemical 5-pGpGsodIum salt 700ug
SDP
Catalog No. 502951036
Encompass_Preferred
A linearized form of cyclic di-GMP; intended for use as a negative control for cyclic di-GMP signaling
Catalog No. 50-295-1036 Supplier Cayman Chemical Supplier No. 33889700 UG
A linearized form of cyclic di-GMP; intended for use as a negative control for cyclic di-GMP signaling