Learn More
Medchemexpress LLC HY-134653 10mg , 5-Ph-IAA CAS:168649-23-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13465310MG
Encompass_Preferred
HY-134653 10mg 5-Ph-IAA CAS: 168649-23-8 5-Ph-IAA is a derivative of IAA. 5-Ph-IAA, a ligand, establishes the auxin-inducible degron 2 (AID2) system together with an OsTIR1 (F74G) mutant. AID2 induces rapid and efficient depletion of mAID-fused proteins to study protein function in living cells, causing tumor suppression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8953